Assay Details
| Gene Symbol: Aadacl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128091.1 | NP_001121563.1 | 3 - 4 | 666 | 87 | |
| GenBank mRNA | BC147724.1 | - | 3 - 4 | 666 | 9056673 | 87 |
| BC147728.1 | - | 3 - 4 | 666 | 9056677 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aadacl2
Genomic Map