Assay Details
| Gene Symbol: Alpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081082.2 | NP_001074551.1 | 5 - 6 | 725 | 110 | |
| GenBank mRNA | BC119800.1 | - | 5 - 6 | 653 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Alpi
Genomic Map