Assay Details
| Gene Symbol: Cntn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177129.5 | NP_796103.2 | 1 - 2 | 215 | 69 | |
| GenBank mRNA | AK147573.1 | - | 1 - 2 | 141 | 69 | |
| AK147602.1 | - | 1 - 2 | 216 | 69 | ||
| AK147658.1 | - | 1 - 2 | 141 | 69 | ||
| AK147673.1 | - | 1 - 2 | 166 | 69 | ||
| BC066106.1 | - | 1 - 2 | 135 | 30362931 | 69 | |
| CJ098637.1 | - | 1 - 2 | 216 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK147602.1, AK147673.1, BC066106.1, NM_177129.5 transcripts.
Set Membership: |
|

Cntn2
Genomic Map