Assay Details
| Gene Symbol: Nuak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028778.4 | NP_083054.1 | 2 - 3 | 460 | 95 | |
| GenBank mRNA | AK004737.2 | - | 2 - 3 | 448 | 95 | |
| AK033672.1 | - | 2 - 3 | 447 | 95 | ||
| BC033302.1 | - | 2 - 3 | 435 | 3663595 | 95 | |
More Information
Set Membership: |
|

Nuak2
Genomic Map