| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100022956.mm.1 |
NM_080850.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022956.mm.1 |
NM_080850.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022956.mm.1 |
TC0100002832.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100022955.mm.1 |
NM_080850.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022955.mm.1 |
NM_080850.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022955.mm.1 |
TC0100002832.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100022957.mm.1 |
NM_080850.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022957.mm.1 |
NM_080850.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022957.mm.1 |
TC0100002832.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100022958.mm.1 |
NM_080850.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022958.mm.1 |
NM_080850.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022958.mm.1 |
TC0100002832.mm.1 |
MTA TC |