Assay Details
| Gene Symbol: Crocc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK086911.1 | - | 5 - 6 | 717 | 90 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P876147 | AK086911.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P876147 | AK086911.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 457943 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | AK076403.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | AK159232.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | AK160828.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | AK160879.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | AK161554.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | AK163368.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | AK166775.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | AK167331.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | AK172048.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | BC012220.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | BC019769.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | BC023074.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | BC023169.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | BC025523.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | BC025557.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | BC029003.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | BC039275.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | BC042622.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | BC056633.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | NM_173363.4 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | NM_178041.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | XM_001479591.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596226 | XM_001479599.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654995 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 252953 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 362355 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795768 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 829534 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 813118 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 713769 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 32917 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 146822 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162118 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151521 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P876147 | AK086911.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P876147 | AK086911.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162118 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P151521 | - | - |
| Whole Mouse Genome G4122A | A_52_P876147 | AK086911.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P876147 | AK086911.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162118 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P151521 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00092599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00092627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00771217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454019 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00375927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454146 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00092589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454060 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454046 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00092637 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454054 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00092553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454077 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454002 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00092584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454025 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454075 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00123500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00092560 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454040 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454036 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454017 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00375902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454143 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00375906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00092662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454007 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454035 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00092626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454064 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00205486 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454005 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454073 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00092612 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00092668 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20454042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00092555 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10348718 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348725 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348723 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348734 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348722 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348716 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348737 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348721 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348717 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348719 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348730 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348714 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348731 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348732 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348738 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348720 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348715 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348727 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348733 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348736 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348735 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348728 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348726 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348729 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348724 | - | - |

Crocc2
Genomic Map