| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100003657.mm.1 |
NM_001163357.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003657.mm.1 |
NM_001163357.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003657.mm.1 |
TC1100000477.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100003658.mm.1 |
NM_001163357.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003658.mm.1 |
NM_001163357.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003658.mm.1 |
TC1100000477.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100003656.mm.1 |
NM_001163357.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003656.mm.1 |
NM_001163357.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003656.mm.1 |
TC1100000477.mm.1 |
MTA TC |