Assay Details
Gene Symbol: Gm12169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163357.1 | NP_001156829.1 | 4 - 5 | 557 | 103 | |
GenBank mRNA | BG066735.2 | - | 3 - 4 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
