Assay Details
| Gene Symbol: Itk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281965.1 | NP_001268894.1 | 3 - 4 | 452 | 91 | |
| NM_001281966.1 | NP_001268895.1 | 3 - 4 | 434 | 91 | ||
| NM_001281967.1 | NP_001268896.1 | 3 - 4 | 434 | 91 | ||
| NM_001281968.1 | NP_001268897.1 | 3 - 4 | 431 | 91 | ||
| NM_010583.3 | NP_034713.2 | 3 - 4 | 434 | 91 | ||
| GenBank mRNA | AK020772.1 | - | 3 - 4 | 382 | 91 | |
| AK079693.1 | - | 3 - 4 | 382 | 91 | ||
| AK083380.1 | - | 3 - 4 | 405 | 91 | ||
| AK153692.1 | - | 3 - 4 | 381 | 91 | ||
| BC028478.1 | - | 3 - 4 | 376 | 91 | ||
| BC128374.1 | - | 3 - 4 | 360 | 40111687 | 91 | |
| BC128375.1 | - | 3 - 4 | 357 | 40111688 | 91 | |
| D14042.1 | - | 3 - 4 | 374 | 91 | ||
| L00619.1 | - | 3 - 4 | 439 | 91 | ||
| L05631.1 | - | 3 - 4 | 400 | 91 | ||
| L10628.1 | - | 3 - 4 | 402 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itk
Genomic Map