Assay Details
| Gene Symbol: Prlh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101647.1 | NP_001095117.1 | 1 - 2 | 104 | 58 | |
| GenBank mRNA | FJ424613.1 | - | 1 - 2 | 104 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
tissue homeostasis
response to dietary excess
reduction of food intake in response to dietary excess
energy reserve metabolic process
lipid metabolic process
G-protein coupled receptor signaling pathway
feeding behavior
response to glucose
response to insulin
regulation of multicellular organism growth
eating behavior
response to peptide hormone
fat cell differentiation
autonomic nervous system development
neuropeptide hormone activity
prolactin-releasing peptide receptor binding
tissue homeostasis
response to dietary excess
reduction of food intake in response to dietary excess
energy reserve metabolic process
lipid metabolic process
G-protein coupled receptor signaling pathway
feeding behavior
response to glucose
response to insulin
regulation of multicellular organism growth
eating behavior
response to peptide hormone
fat cell differentiation
autonomic nervous system development
neuropeptide hormone activity
prolactin-releasing peptide receptor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P111049 | NM_001101647.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P111049 | FJ424613.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 208398 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P111049 | NM_001101647.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P111049 | FJ424613.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20439486 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00224022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20439474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20439465 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20439469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20439481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20439463 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20439452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20439488 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20439471 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20439459 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20439473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20439464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20439461 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00364649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20439476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00447425 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20439453 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20439468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00447420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20439457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00447426 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20439445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00224021 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00364652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20439470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20439451 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00799636 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00799634 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10348490 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348492 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10348489 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100007442.mm.1 | NM_001101647.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100007442.mm.1 | NM_001101647.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100007442.mm.1 | TC0100000883.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100007441.mm.1 | NM_001101647.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100007441.mm.1 | NM_001101647.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100007441.mm.1 | TC0100000883.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 734562 | - | - |

Prlh
Genomic Map