Assay Details
| Gene Symbol: Rgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015811.2 | NP_056626.2 | 2 - 3 | 285 | 66 | |
| GenBank mRNA | AF215667.1 | - | 2 - 3 | 216 | 66 | |
| AK138688.1 | - | 2 - 3 | 241 | 66 | ||
| AK170366.1 | - | 2 - 3 | 237 | 66 | ||
| BC028634.1 | - | 2 - 3 | 216 | 1328269 | 66 | |
| BU695313.1 | - | 2 - 3 | 307 | 66 | ||
More Information
Set Membership: |
|

Rgs1
Genomic Map