Assay Details
| Gene Symbol: Rgs18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022881.4 | NP_075019.1 | 1 - 2 | 298 | 81 | |
| GenBank mRNA | AB042807.1 | - | 1 - 2 | 313 | 81 | |
| AF302685.1 | - | 1 - 2 | 304 | 81 | ||
| AK036407.1 | - | 1 - 2 | 299 | 81 | ||
| BC099387.1 | - | 1 - 2 | 293 | 30846527 | 81 | |
| BC132544.1 | - | 1 - 2 | 299 | 40130821 | 81 | |
| BC132546.1 | - | 1 - 2 | 299 | 40130823 | 81 | |
More Information
Set Membership: |
|

Rgs18
Genomic Map