Assay Details
| Gene Symbol: Dbi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037999.2 | NP_001033088.1 | 3 - 4 | 339 | 63 | |
| NM_007830.4 | NP_031856.1 | 3 - 4 | 333 | 63 | ||
| GenBank mRNA | AA068307.1 | - | 3 - 4 | 279 | 63 | |
| AK008540.1 | - | 3 - 4 | 254 | 63 | ||
| AK008576.1 | - | 3 - 4 | 254 | 63 | ||
| AK018720.1 | - | 3 - 4 | 255 | 63 | ||
| AK020692.1 | - | 3 - 4 | 303 | 63 | ||
| AK027906.1 | - | 3 - 4 | 255 | 63 | ||
| AK078428.1 | - | 3 - 4 | 317 | 63 | ||
| AK145278.1 | - | 3 - 4 | 255 | 63 | ||
| AK148753.1 | - | 3 - 4 | 255 | 63 | ||
| AM000002.1 | - | 3 - 4 | 339 | 63 | ||
| AV001592.1 | - | 3 - 4 | 227 | 63 | ||
| BC028874.1 | - | 3 - 4 | 233 | 4194069 | 63 | |
| CJ183794.1 | - | 3 - 4 | 334 | 63 | ||
| EF437365.1 | - | 3 - 4 | 240 | 63 | ||
| X61431.1 | - | 3 - 4 | 252 | 63 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
behavioral fear response
hair follicle development
triglyceride metabolic process
transport
learning or memory
glial cell proliferation
lateral ventricle development
negative regulation of fatty acid beta-oxidation
regulation of synaptic transmission, GABAergic
phosphatidylcholine acyl-chain remodeling
skin development
positive regulation of lipid biosynthetic process
positive regulation of release of sequestered calcium ion into cytosol
long-term synaptic potentiation
fatty-acyl-CoA binding
lipid binding
benzodiazepine receptor binding
long-chain fatty acyl-CoA binding
protein dimerization activity
behavioral fear response
hair follicle development
triglyceride metabolic process
transport
learning or memory
glial cell proliferation
lateral ventricle development
negative regulation of fatty acid beta-oxidation
regulation of synaptic transmission, GABAergic
phosphatidylcholine acyl-chain remodeling
skin development
positive regulation of lipid biosynthetic process
positive regulation of release of sequestered calcium ion into cytosol
long-term synaptic potentiation
fatty-acyl-CoA binding
lipid binding
benzodiazepine receptor binding
long-chain fatty acyl-CoA binding
protein dimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20560374 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20560372 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20560373 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2049979 | AK027906.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2049979 | AK145278.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2049979 | AV001592.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2049979 | BC028874.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2049979 | NM_001037999.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2116924 | AK008576.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2116924 | AK018720.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2116924 | AK020692.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2116924 | AK027906.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2116924 | AK145278.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 361309 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 805669 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 44273 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 41439 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158124 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2116924 | AK008576.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2116924 | AK018720.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2116924 | AK020692.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2116924 | AK027906.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2116924 | AK145278.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2049979 | AK027906.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2049979 | AK145278.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2049979 | AV001592.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2049979 | BC028874.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2049979 | NM_001037999.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P399697 | X61431.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P399697 | AK008576.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P399697 | AK018720.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P399697 | AK020692.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P399697 | AK027906.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560365 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560372 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560350 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20560369 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04239240 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158124 | - | - |
| Whole Mouse Genome G4122A | A_51_P399697 | X61431.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P399697 | AK008576.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P399697 | AK018720.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P399697 | AK020692.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P399697 | AK027906.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115180 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158124 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00867352 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00867350 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00867363 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00867373 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20560372 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20560374 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20560373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20560362 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00299574 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20560332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20560369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20560367 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00867353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00570096 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20560353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20560372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00570095 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20560362 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20560351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20560336 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20560364 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20560338 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20560339 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00867358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20560340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20560374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20560334 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20560365 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20560350 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20560345 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00830464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20560358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00302038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00423614 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20560344 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20560352 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00423611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20560371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20560366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00302035 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10357244 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376095 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357248 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357246 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357245 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357243 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357247 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433991_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438093_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422432_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455976_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10376094 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10357242 | - | - |
| Mouse Genome 430 2.0 Array | 1433991_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1455976_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1422432_at | - | - |
| Mouse Genome 430 2.0 Array | 1422432_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438093_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1433991_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1455976_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1438093_PM_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13167_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13167_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100023838.mm.1 | NM_001037999.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023838.mm.1 | NM_001037999.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023838.mm.1 | NM_007830.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023838.mm.1 | NM_007830.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023838.mm.1 | TC0100002964.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100023840.mm.1 | NM_001037999.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023840.mm.1 | NM_001037999.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023840.mm.1 | NM_007830.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023840.mm.1 | NM_007830.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023840.mm.1 | TC0100002964.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100023839.mm.1 | NM_001037999.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023839.mm.1 | NM_001037999.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023839.mm.1 | NM_007830.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023839.mm.1 | NM_007830.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023839.mm.1 | TC0100002964.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100023841.mm.1 | NM_001037999.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023841.mm.1 | NM_001037999.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023841.mm.1 | NM_007830.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023841.mm.1 | NM_007830.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023841.mm.1 | TC0100002964.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455976_x_at | - | - |
| Mouse Expression Set 430 | 1438093_PM_x_at | - | - |
| Mouse Expression Set 430 | 1422432_PM_at | - | - |
| Mouse Expression Set 430 | 1422432_at | - | - |
| Mouse Expression Set 430 | 1433991_x_at | - | - |
| Mouse Expression Set 430 | 1438093_x_at | - | - |
| Mouse Expression Set 430 | 1455976_PM_x_at | - | - |
| Mouse Expression Set 430 | 1433991_PM_x_at | - | - |
| Murine Genome U74v2 Set | 97248_at | - | - |
| Murine Genome U74v2 Set | 97248_PM_at | - | - |
| Murine Genome U74v2 Set | 169069_i_at | - | - |
| Murine Genome U74v2 Set | 169069_PM_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422432_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455976_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438093_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422432_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455976_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433991_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433991_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438093_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433991_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433991_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438093_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455976_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438093_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422432_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422432_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455976_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 611934 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1090706 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1090706 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001090706 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1215469 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215469 | - | - |
| mouse-6 v1.1 (Illumina) | scl16375.7_22-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1090706 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl16375.7_22-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1090706 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1215469 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3150159 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3071356 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3150159 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3071356 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1215469 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl16375.7_22-S | - | - |

Dbi
Genomic Map