Assay Details
| Gene Symbol: 4933405E24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045506.1 | 1 - 2 | 452 | 98 | ||
| GenBank mRNA | AK016664.1 | - | 1 - 2 | 455 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

4933405E24Rik
Genomic Map