Assay Details
| Gene Symbol: Egln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028133.2 | NP_082409.2 | 3 - 4 | 925 | 71 | |
| GenBank mRNA | AJ310548.1 | - | 3 - 4 | 906 | 71 | |
| AK011502.1 | - | 2 - 3 | 229 | 71 | ||
| AK044787.1 | - | 3 - 4 | 927 | 71 | ||
| AK165972.1 | - | 3 - 4 | 856 | 71 | ||
| AK170732.1 | - | 3 - 4 | 856 | 71 | ||
| BC044926.1 | - | 3 - 4 | 835 | 5373240 | 71 | |
| BC058278.1 | - | 3 - 4 | 845 | 6528802 | 71 | |
| BC069893.1 | - | 3 - 4 | 853 | 4978765 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Egln3
Genomic Map