Assay Details
| Gene Symbol: 4930512B01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033573.1 | 7 - 8 | 983 | 123 | ||
| GenBank mRNA | AK015558.1 | - | 7 - 8 | 993 | 123 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitogen-activated protein kinase kinase kinase kinase 5 | Map4k5 |
4432415E19Rik, GCKR, KHS, MAPKKKK5 |
Anti-sense | Chr. 12 - 69803757 - 69893163 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4931403G20 gene | 4931403G20Rik | 1.8kb | Sense | Chr. 12 - 69843686 - 69853671 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930512B01Rik
Genomic Map