Assay Details
| Gene Symbol: Zfp354a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009329.3 | NP_033355.2 | 4 - 5 | 457 | 84 | |
| GenBank mRNA | AB221586.1 | - | 3 - 4 | 261 | 84 | |
| AF184111.1 | - | 4 - 5 | 419 | 84 | ||
| AK030469.1 | - | 4 - 5 | 436 | 84 | ||
| BC050843.1 | - | 4 - 5 | 457 | 6506256 | 84 | |
| BC087540.1 | - | 4 - 5 | 593 | 4239789 | 84 | |
More Information
Set Membership: |
|

Zfp354a
Genomic Map