Assay Details
| Gene Symbol: Rsad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021384.4 | NP_067359.2 | 1 - 2 | 380 | 77 | |
| GenBank mRNA | AF236064.2 | - | 1 - 2 | 356 | 77 | |
| AF442152.1 | - | 1 - 2 | 341 | 77 | ||
| AK010906.1 | - | 1 - 2 | 360 | 77 | ||
| AK036373.1 | - | 1 - 2 | 360 | 77 | ||
| AK149900.1 | - | 1 - 2 | 362 | 77 | ||
| AK150069.1 | - | 1 - 2 | 362 | 77 | ||
| AK150347.1 | - | 1 - 2 | 362 | 77 | ||
| AK150406.1 | - | 1 - 2 | 360 | 77 | ||
| AK150425.1 | - | 1 - 2 | 362 | 77 | ||
| AK150525.1 | - | 1 - 2 | 361 | 77 | ||
| AK150536.1 | - | 1 - 2 | 362 | 77 | ||
| AK150616.1 | - | 1 - 2 | 360 | 77 | ||
| AK150684.1 | - | 1 - 2 | 362 | 77 | ||
| AK150731.1 | - | 1 - 2 | 362 | 77 | ||
| AK150767.1 | - | 1 - 2 | 362 | 77 | ||
| AK150783.1 | - | 1 - 2 | 362 | 77 | ||
| AK150788.1 | - | 1 - 2 | 362 | 77 | ||
| AK150812.1 | - | 1 - 2 | 362 | 77 | ||
| AK150835.1 | - | 1 - 2 | 361 | 77 | ||
| AK151140.1 | - | 1 - 2 | 362 | 77 | ||
| AK151185.1 | - | 1 - 2 | 362 | 77 | ||
| AK151191.1 | - | 1 - 2 | 362 | 77 | ||
| AK151206.1 | - | 1 - 2 | 362 | 77 | ||
| AK151286.1 | - | 1 - 2 | 360 | 77 | ||
| AK151308.1 | - | 1 - 2 | 362 | 77 | ||
| AK151331.1 | - | 1 - 2 | 362 | 77 | ||
| AK151339.1 | - | 1 - 2 | 360 | 77 | ||
| AK151389.1 | - | 1 - 2 | 362 | 77 | ||
| AK151410.1 | - | 1 - 2 | 361 | 77 | ||
| AK151412.1 | - | 1 - 2 | 360 | 77 | ||
| AK151414.1 | - | 1 - 2 | 361 | 77 | ||
| AK151476.1 | - | 1 - 2 | 362 | 77 | ||
| AK151502.1 | - | 1 - 2 | 362 | 77 | ||
| AK151538.1 | - | 1 - 2 | 362 | 77 | ||
| AK151767.1 | - | 1 - 2 | 362 | 77 | ||
| AK151770.1 | - | 1 - 2 | 360 | 77 | ||
| AK151825.1 | - | 1 - 2 | 360 | 77 | ||
| AK151833.1 | - | 1 - 2 | 360 | 77 | ||
| AK151846.1 | - | 1 - 2 | 360 | 77 | ||
| AK151906.1 | - | 1 - 2 | 362 | 77 | ||
| AK151909.1 | - | 1 - 2 | 360 | 77 | ||
| AK151940.1 | - | 1 - 2 | 360 | 77 | ||
| AK151954.1 | - | 1 - 2 | 362 | 77 | ||
| AK151971.1 | - | 1 - 2 | 362 | 77 | ||
| AK152050.1 | - | 1 - 2 | 360 | 77 | ||
| AK152065.1 | - | 1 - 2 | 362 | 77 | ||
| AK152075.1 | - | 1 - 2 | 362 | 77 | ||
| AK152235.1 | - | 1 - 2 | 360 | 77 | ||
| AK152239.1 | - | 1 - 2 | 362 | 77 | ||
| AK152241.1 | - | 1 - 2 | 362 | 77 | ||
| AK152310.1 | - | 1 - 2 | 360 | 77 | ||
| AK152354.1 | - | 1 - 2 | 360 | 77 | ||
| AK152493.1 | - | 1 - 2 | 362 | 77 | ||
| AK152600.1 | - | 1 - 2 | 360 | 77 | ||
| AK152601.1 | - | 1 - 2 | 362 | 77 | ||
| AK152631.1 | - | 1 - 2 | 360 | 77 | ||
| AK152644.1 | - | 1 - 2 | 362 | 77 | ||
| AK152664.1 | - | 1 - 2 | 362 | 77 | ||
| AK152701.1 | - | 1 - 2 | 362 | 77 | ||
| AK152796.1 | - | 1 - 2 | 360 | 77 | ||
| AK152861.1 | - | 1 - 2 | 360 | 77 | ||
| AK152880.1 | - | 1 - 2 | 360 | 77 | ||
| AK152895.1 | - | 1 - 2 | 362 | 77 | ||
| AK152934.1 | - | 1 - 2 | 362 | 77 | ||
| AK152962.1 | - | 1 - 2 | 360 | 77 | ||
| AK153003.1 | - | 1 - 2 | 362 | 77 | ||
| AK153024.1 | - | 1 - 2 | 362 | 77 | ||
| AK153062.1 | - | 1 - 2 | 364 | 77 | ||
| AK153077.1 | - | 1 - 2 | 360 | 77 | ||
| AK153078.1 | - | 1 - 2 | 360 | 77 | ||
| AK153232.1 | - | 1 - 2 | 362 | 77 | ||
| AK153234.1 | - | 1 - 2 | 362 | 77 | ||
| AK153306.1 | - | 1 - 2 | 360 | 77 | ||
| AK153318.1 | - | 1 - 2 | 362 | 77 | ||
| AK153324.1 | - | 1 - 2 | 362 | 77 | ||
| AK153350.1 | - | 1 - 2 | 362 | 77 | ||
| AK153435.1 | - | 1 - 2 | 238 | 77 | ||
| AK153556.1 | - | 1 - 2 | 362 | 77 | ||
| AK159147.1 | - | 1 - 2 | 360 | 77 | ||
| BC057868.1 | - | 1 - 2 | 353 | 5327504 | 77 | |
| CK623585.1 | - | 1 - 2 | 380 | 77 | ||
| CT010378.1 | - | 1 - 2 | 347 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK150069.1, AK151331.1 transcripts.
Set Membership: |
|

Rsad2
Genomic Map