Assay Details
| Gene Symbol: Rnf144a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080563.4 | NP_542130.1 | 1 - 2 | 165 | 84 | |
| GenBank mRNA | AK038586.1 | - | 1 - 2 | 57 | 84 | |
| BC030187.1 | - | 1 - 2 | 129 | 5345434 | 84 | |
| BQ884505.1 | - | 1 - 2 | 98 | 84 | ||
| BY753828.1 | - | 1 - 2 | 159 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK038586.1, BC030187.1, NM_080563.4 transcripts.
Set Membership: |
|

Rnf144a
Genomic Map