Assay Details
| Gene Symbol: Eif4e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039169.1 | NP_001034258.1 | 4 - 5 | 909 | 95 | |
| NM_001039170.1 | NP_001034259.1 | 4 - 5 | 458 | 95 | ||
| NM_023314.3 | NP_075803.2 | 4 - 5 | 458 | 95 | ||
| GenBank mRNA | AF068116.1 | - | 4 - 5 | 372 | 95 | |
| AK017221.1 | - | 4 - 5 | 910 | 95 | ||
| AK018516.1 | - | 4 - 5 | 456 | 95 | ||
| AK032965.1 | - | 4 - 5 | 462 | 95 | ||
| AK076067.1 | - | 2 - 3 | 173 | 95 | ||
| AK138055.1 | - | 4 - 5 | 397 | 95 | ||
| AK150495.1 | - | 4 - 5 | 397 | 95 | ||
| BC028903.1 | - | 4 - 5 | 508 | 4166275 | 95 | |
| BC045153.1 | - | 4 - 5 | 392 | 6441327 | 95 | |
| BC049077.1 | - | 4 - 5 | 426 | 5708556 | 95 | |
| U01137.1 | - | 4 - 5 | 95 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Eif4e2
Genomic Map