Assay Details
| Gene Symbol: Gm34388 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK054506.1 | - | 2 - 3 | 388 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 5330416C01 gene | 5330416C01Rik | Gm532 | -86.9kb | Sense | Chr. 13 - 97712315 - 97778952 on NCBI Build GRCm38 |
| Downstream | Rho guanine nucleotide exchange factor (GEF) 28 | Arhgef28 |
9230110L08Rik, AI323540, D13Bwg1089e, RIP2, Rgnef, RhoGEF, Rhoip2, p190RhoGEF |
22.5kb | Sense | Chr. 13 - 97898594 - 98206624 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm34388
Genomic Map