Assay Details
| Gene Symbol: Fmo9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172844.2 | NP_766432.1 | 3 - 4 | 342 | 99 | |
| GenBank mRNA | AK029219.1 | - | 3 - 4 | 343 | 99 | |
| AK161224.1 | - | 3 - 4 | 342 | 99 | ||
| BC112405.1 | - | 3 - 4 | 332 | 40060428 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fmo9
Genomic Map