Assay Details
| Gene Symbol: Pag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195031.1 | NP_001181960.1 | 2 - 3 | 529 | 75 | |
| GenBank mRNA | AK040230.1 | - | 2 - 3 | 479 | 75 | |
| AK156000.1 | - | 2 - 3 | 481 | 75 | ||
| AK156986.1 | - | 2 - 3 | 454 | 75 | ||
| AK172106.1 | - | 2 - 3 | 452 | 75 | ||
| BC145328.1 | - | 2 - 3 | 353 | 9053884 | 75 | |
| BC145761.1 | - | 2 - 3 | 353 | 40131064 | 75 | |
| BC145763.1 | - | 2 - 3 | 353 | 40131066 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK040230.1, AK156000.1, AK172106.1, BC145328.1, BC145761.1, BC145763.1, NM_001195031.1 transcripts.
Set Membership: |
|

Pag1
Genomic Map