Assay Details
| Gene Symbol: 4930577N17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073429.1 | 1 | 1480 | 149 | ||
| GenBank mRNA | AK017883.1 | - | 1 | 1349 | 149 | |
| AK086057.1 | - | 1 | 1425 | 149 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CCR4 carbon catabolite repression 4-like (S. cerevisiae) | Ccrn4l |
AU043840, Ccr4, nocturnin |
-25.2kb | Sense | Chr. 3 - 51224447 - 51251654 on NCBI Build GRCm38 |
| Downstream | predicted gene 5103 | Gm5103 | A630062H14 | 99.7kb | Sense | Chr. 3 - 51378025 - 51386324 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930577N17Rik
Genomic Map