Assay Details
| Gene Symbol: Usp9x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009481.2 | NP_033507.2 | 18 - 19 | 3029 | 143 | |
| GenBank mRNA | DQ086491.1 | - | 18 - 19 | 2990 | 143 | |
| U67874.1 | - | 18 - 19 | 2990 | 143 | ||
| Gene Symbol: Gm40399 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK138386.1 | - | 2 | 959 | 143 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Usp9x
Genomic Map