Assay Details
| Gene Symbol: Fcamr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170632.1 | NP_001164103.1 | 2 - 3 | 310 | 93 | |
| NM_144960.2 | NP_659209.2 | 2 - 3 | 142 | 93 | ||
| GenBank mRNA | BC110550.1 | - | 2 - 3 | 145 | 40047153 | 93 |
| FJ011703.1 | - | 1 - 2 | 223 | 93 | ||
More Information
Set Membership: |
|

Fcamr
Genomic Map