Assay Details
| Gene Symbol: 1700111N16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033213.2 | 7 - 8 | 657 | 101 | ||
| GenBank mRNA | AK007172.1 | - | 7 - 8 | 658 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

1700111N16Rik
Genomic Map