Assay Details
| Gene Symbol: Impa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018864.5 | NP_061352.2 | 2 - 3 | 188 | 80 | |
| GenBank mRNA | AF042730.1 | - | 2 - 3 | 133 | 80 | |
| AK011770.1 | - | 2 - 3 | 126 | 80 | ||
| AK078406.1 | - | 2 - 3 | 137 | 80 | ||
| AK083238.1 | - | 2 - 3 | 122 | 80 | ||
| AK145472.1 | - | 125 | 80 | |||
| AK147778.1 | - | 2 - 3 | 148 | 80 | ||
| AK152774.1 | - | 2 - 3 | 139 | 80 | ||
| AK153457.1 | - | 2 - 3 | 155 | 80 | ||
| AK160203.1 | - | 2 - 3 | 126 | 80 | ||
| AK160467.1 | - | 2 - 3 | 120 | 80 | ||
| AK165977.1 | - | 2 - 3 | 130 | 80 | ||
| AK168457.1 | - | 2 - 3 | 163 | 80 | ||
| BC049080.1 | - | 2 - 3 | 129 | 6419835 | 80 | |
| BC055722.1 | - | 1 - 2 | 83 | 6413389 | 80 | |
| BY283562.1 | - | 2 - 3 | 189 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Impa1
Genomic Map