Assay Details
| Gene Symbol: Impa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018864.5 | NP_061352.2 | 5 - 6 | 467 | 68 | |
| GenBank mRNA | AK011770.1 | - | 5 - 6 | 405 | 68 | |
| AK078406.1 | - | 5 - 6 | 416 | 68 | ||
| AK083238.1 | - | 5 - 6 | 401 | 68 | ||
| AK145472.1 | - | 404 | 68 | |||
| AK147778.1 | - | 5 - 6 | 427 | 68 | ||
| AK152774.1 | - | 5 - 6 | 418 | 68 | ||
| AK160203.1 | - | 5 - 6 | 405 | 68 | ||
| AK160467.1 | - | 5 - 6 | 399 | 68 | ||
| AK165977.1 | - | 5 - 6 | 409 | 68 | ||
| AK168457.1 | - | 5 - 6 | 442 | 68 | ||
| BC049080.1 | - | 5 - 6 | 408 | 6419835 | 68 | |
| BC055722.1 | - | 4 - 5 | 362 | 6413389 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Impa1
Genomic Map