Assay Details
| Gene Symbol: Atp6v0a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243049.1 | NP_001229978.1 | 10 - 11 | 1179 | 85 | |
| NM_001243050.1 | NP_001229979.1 | 11 - 12 | 1357 | 85 | ||
| NM_001243051.1 | NP_001229980.1 | 11 - 12 | 1378 | 85 | ||
| NM_016920.3 | NP_058616.1 | 11 - 12 | 1357 | 85 | ||
| GenBank mRNA | AB022321.1 | - | 10 - 11 | 1186 | 85 | |
| AF218249.1 | - | 11 - 12 | 1327 | 85 | ||
| AF218250.1 | - | 10 - 11 | 1201 | 85 | ||
| AF218251.1 | - | 10 - 11 | 1180 | 85 | ||
| AK029316.1 | - | 10 - 11 | 3111 | 85 | ||
| AK158786.1 | - | 10 - 11 | 1147 | 85 | ||
| AK159114.1 | - | 11 - 12 | 1293 | 85 | ||
| AK192645.1 | - | 1 - 2 | 99 | 85 | ||
| AK196696.1 | - | 1 - 2 | 100 | 85 | ||
| AK198740.1 | - | 1 - 2 | 99 | 85 | ||
| AK201730.1 | - | 1 - 2 | 99 | 85 | ||
| BC066839.1 | - | 11 - 12 | 1322 | 30101340 | 85 | |
| BC071182.1 | - | 11 - 12 | 1322 | 30101184 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v0a1
Genomic Map