Assay Details
| Gene Symbol: Atp6v0a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243049.1 | NP_001229978.1 | 11 - 12 | 1314 | 76 | |
| NM_001243050.1 | NP_001229979.1 | 12 - 13 | 1492 | 76 | ||
| NM_001243051.1 | NP_001229980.1 | 12 - 13 | 1513 | 76 | ||
| NM_016920.3 | NP_058616.1 | 12 - 13 | 1492 | 76 | ||
| GenBank mRNA | AB022321.1 | - | 11 - 12 | 1321 | 76 | |
| AF218249.1 | - | 12 - 13 | 1462 | 76 | ||
| AF218250.1 | - | 11 - 12 | 1336 | 76 | ||
| AF218251.1 | - | 11 - 12 | 1315 | 76 | ||
| AK029316.1 | - | 11 - 12 | 3246 | 76 | ||
| AK158786.1 | - | 11 - 12 | 1282 | 76 | ||
| AK159114.1 | - | 12 - 13 | 1428 | 76 | ||
| AK172078.1 | - | 12 - 13 | 1422 | 76 | ||
| BC066839.1 | - | 12 - 13 | 1457 | 30101340 | 76 | |
| BC071182.1 | - | 12 - 13 | 1457 | 30101184 | 76 | |
| U13836.1 | - | 11 - 12 | 1354 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v0a1
Genomic Map