Assay Details
Gene Symbol: C030034L19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK039918.1 | - | 181 | 99 | ||
AK081263.1 | - | 177 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger and BTB domain containing 10 | Zbtb10 |
4832414A18, AU021107, RINZF |
-117.7kb | Sense | Chr. 3 - 9250567 - 9285333 on NCBI Build GRCm38 |
Downstream | zinc finger protein 704 | Zfp704 |
C030026M03, C030034L19Rik, Gig1, Znf704 |
21.5kb | Anti-sense | Chr. 3 - 9427010 - 9610085 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
