Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300000057.mm.1 |
NM_030708.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000057.mm.1 |
NM_030708.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000057.mm.1 |
TC0300000017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300000062.mm.1 |
NM_030708.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000062.mm.1 |
NM_030708.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000062.mm.1 |
TC0300000017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300000058.mm.1 |
NM_030708.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000058.mm.1 |
NM_030708.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000058.mm.1 |
TC0300000017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300000061.mm.1 |
NM_030708.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000061.mm.1 |
NM_030708.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000061.mm.1 |
TC0300000017.mm.1 |
MTA TC |