Assay Details
Gene Symbol: Zfhx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030708.2 | NP_109633.2 | 4 - 5 | 3663 | 77 | |
GenBank mRNA | AB024499.1 | - | 3 - 4 | 3287 | 77 | |
BI988177.1 | - | 2 - 3 | 366 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
