Assay Details
Gene Symbol: Zfhx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030708.2 | NP_109633.2 | 5 - 6 | 3722 | 69 | |
GenBank mRNA | AB024499.1 | - | 4 - 5 | 3346 | 69 | |
BI988177.1 | - | 3 - 4 | 425 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
