Assay Details
| Gene Symbol: Zfhx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030708.2 | NP_109633.2 | 7 - 8 | 3973 | 102 | |
| GenBank mRNA | AB024499.1 | - | 6 - 7 | 3597 | 102 | |
More Information
Set Membership: |
|

Zfhx4
Genomic Map