Assay Details
| Gene Symbol: Rad17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044371.2 | NP_001037836.1 | 7 - 8 | 903 | 107 | |
| NM_001283011.1 | NP_001269940.1 | 8 - 9 | 1108 | 107 | ||
| NM_011233.3 | NP_035363.2 | 7 - 8 | 1031 | 107 | ||
| GenBank mRNA | AF085737.1 | - | 7 - 8 | 954 | 107 | |
| AJ011923.1 | - | 6 - 7 | 804 | 107 | ||
| AK134277.1 | - | 7 - 8 | 858 | 107 | ||
| AK162016.1 | - | 7 - 8 | 890 | 107 | ||
| AK167358.1 | - | 8 - 9 | 1051 | 107 | ||
| BC023939.1 | - | 7 - 8 | 908 | 107 | ||
| BC050267.1 | - | 4 - 5 | 543 | 107 | ||
| BC066855.1 | - | 7 - 8 | 1021 | 30473984 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rad17
Genomic Map