Assay Details
| Gene Symbol: Ranbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011240.3 | NP_035370.2 | 10 - 11 | 1586 | 126 | |
| GenBank mRNA | AK145306.1 | - | 10 - 11 | 1517 | 126 | |
More Information
Set Membership: |
|

Ranbp2
Genomic Map