Assay Details
| Gene Symbol: Gcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027375.2 | NP_081651.2 | 10 - 11 | 3198 | 127 | |
| GenBank mRNA | AB093232.1 | - | 10 - 11 | 3201 | 127 | |
| AK148283.1 | - | 5 - 6 | 413 | 127 | ||
| BC023811.1 | - | 5 - 6 | 585 | 127 | ||
| BC139007.1 | - | 10 - 11 | 3194 | 8862029 | 127 | |
| BC145495.1 | - | 10 - 11 | 3267 | 127 | ||
| BY741402.1 | - | 5 - 6 | 413 | 127 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gcc2
Genomic Map