Assay Details
| Gene Symbol: Gcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027375.2 | NP_081651.2 | 17 - 18 | 4072 | 85 | |
| GenBank mRNA | AB093232.1 | - | 17 - 18 | 4075 | 85 | |
| AK148283.1 | - | 12 - 13 | 1287 | 85 | ||
| BC023811.1 | - | 12 - 13 | 1459 | 85 | ||
| BC034077.1 | - | 6 - 7 | 710 | 85 | ||
| BC065697.1 | - | 5 - 6 | 467 | 85 | ||
| BC139007.1 | - | 17 - 18 | 4068 | 8862029 | 85 | |
| BC145495.1 | - | 17 - 18 | 4141 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gcc2
Genomic Map