Assay Details
| Gene Symbol: Gcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027375.2 | NP_081651.2 | 21 - 22 | 4811 | 76 | |
| GenBank mRNA | AB093232.1 | - | 21 - 22 | 4814 | 76 | |
| AK148283.1 | - | 16 - 17 | 2026 | 76 | ||
| BC023811.1 | - | 16 - 17 | 2195 | 76 | ||
| BC034077.1 | - | 10 - 11 | 1449 | 76 | ||
| BC065697.1 | - | 9 - 10 | 1206 | 76 | ||
| BC139007.1 | - | 21 - 22 | 4807 | 8862029 | 76 | |
| BC145495.1 | - | 21 - 22 | 4880 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gcc2
Genomic Map