| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000003362.mm.1 |
NM_027375.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003362.mm.1 |
NM_027375.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003362.mm.1 |
TC1000000512.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000003361.mm.1 |
NM_027375.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003361.mm.1 |
NM_027375.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003361.mm.1 |
TC1000000512.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000003363.mm.1 |
NM_027375.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003363.mm.1 |
NM_027375.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003363.mm.1 |
TC1000000512.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000003364.mm.1 |
NM_027375.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003364.mm.1 |
NM_027375.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003364.mm.1 |
TC1000000512.mm.1 |
MTA TC |