Assay Details
| Gene Symbol: Pfkfb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162415.1 | NP_001155887.1 | 5 - 6 | 539 | 136 | |
| NM_001162416.1 | NP_001155888.1 | 3 - 4 | 332 | 136 | ||
| NM_008825.4 | NP_032851.2 | 4 - 5 | 433 | 136 | ||
| NR_027859.1 | 4 - 5 | 433 | 136 | |||
| GenBank mRNA | AK029659.1 | - | 5 - 6 | 540 | 136 | |
| AK146744.1 | - | 4 - 5 | 427 | 136 | ||
| BC018418.1 | - | 4 - 5 | 390 | 3979400 | 136 | |
| BC051014.1 | - | 3 - 4 | 346 | 6493944 | 136 | |
| EU616666.1 | - | 3 - 4 | 331 | 136 | ||
| EU616667.1 | - | 3 - 4 | 331 | 136 | ||
| EU616668.1 | - | 3 - 4 | 331 | 136 | ||
| EU616669.1 | - | 3 - 4 | 331 | 136 | ||
| EU616670.1 | - | 3 - 4 | 341 | 136 | ||
| X98847.1 | - | 3 - 4 | 351 | 136 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the EU616670.1 transcripts.
Set Membership: |
|

Pfkfb2
Genomic Map