Assay Details
| Gene Symbol: Tnip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199275.2 | NP_001186204.1 | 12 - 13 | 1478 | 65 | |
| NM_001199276.2 | NP_001186205.1 | 10 - 11 | 1148 | 65 | ||
| NM_001271455.1 | NP_001258384.1 | 11 - 12 | 1201 | 65 | ||
| NM_001271456.1 | NP_001258385.1 | 12 - 13 | 1373 | 65 | ||
| NM_021327.4 | NP_067302.2 | 11 - 12 | 1320 | 65 | ||
| GenBank mRNA | AJ242777.1 | - | 10 - 11 | 1120 | 65 | |
| AJ242778.1 | - | 11 - 12 | 1292 | 65 | ||
| AK156131.1 | - | 11 - 12 | 1174 | 65 | ||
| AK170841.1 | - | 10 - 11 | 1126 | 65 | ||
| AK172890.1 | - | 1106 | 65 | |||
| AK188523.1 | - | 2 - 3 | 165 | 65 | ||
| AK195559.1 | - | 1 - 2 | 45 | 65 | ||
| AY012159.2 | - | 8 - 9 | 890 | 65 | ||
| BC008186.1 | - | 11 - 12 | 1237 | 3589311 | 65 | |
| BC008665.1 | - | 12 - 13 | 1344 | 3598934 | 65 | |
| CB599739.1 | - | 2 - 3 | 207 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tnip1
Genomic Map