Assay Details
| Gene Symbol: Tnip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199275.2 | NP_001186204.1 | 5 - 6 | 702 | 64 | |
| NM_001199276.2 | NP_001186205.1 | 3 - 4 | 372 | 64 | ||
| NM_001271455.1 | NP_001258384.1 | 4 - 5 | 425 | 64 | ||
| NM_001271456.1 | NP_001258385.1 | 5 - 6 | 597 | 64 | ||
| NM_021327.4 | NP_067302.2 | 4 - 5 | 544 | 64 | ||
| GenBank mRNA | AJ242777.1 | - | 3 - 4 | 344 | 64 | |
| AJ242778.1 | - | 4 - 5 | 516 | 64 | ||
| AK135338.1 | - | 4 - 5 | 523 | 64 | ||
| AK170841.1 | - | 3 - 4 | 350 | 64 | ||
| AK172890.1 | - | 330 | 64 | |||
| AK199662.1 | - | 1 - 2 | 65 | 64 | ||
| AK199664.1 | - | 1 - 2 | 65 | 64 | ||
| AY012159.2 | - | 1 - 2 | 114 | 64 | ||
| BC008186.1 | - | 4 - 5 | 461 | 3589311 | 64 | |
| BC008665.1 | - | 5 - 6 | 568 | 3598934 | 64 | |
| CJ175540.1 | - | 3 - 4 | 367 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tnip1
Genomic Map