Assay Details
| Gene Symbol: Hint1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008248.2 | NP_032274.1 | 1 - 2 | 164 | 84 | |
| GenBank mRNA | AK002965.2 | - | 1 - 2 | 164 | 84 | |
| AK012433.1 | - | 1 - 2 | 170 | 84 | ||
| BC070415.1 | - | 1 - 2 | 209 | 5714321 | 84 | |
| BC080296.1 | - | 1 - 2 | 148 | 6814185 | 84 | |
| U60001.1 | - | 1 - 2 | 134 | 84 | ||
More Information
Set Membership: |
|

Hint1
Genomic Map