Assay Details
| Gene Symbol: Cog6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026225.3 | NP_080501.2 | 4 - 5 | 450 | 118 | |
| GenBank mRNA | AB093286.1 | - | 4 - 5 | 422 | 118 | |
| AK016663.1 | - | 4 - 5 | 463 | 118 | ||
| AK031149.1 | - | 4 - 5 | 444 | 118 | ||
| AK045223.1 | - | 4 - 5 | 444 | 118 | ||
| AK048986.1 | - | 4 - 5 | 437 | 118 | ||
| AK153320.1 | - | 4 - 5 | 439 | 118 | ||
| BC025427.1 | - | 4 - 5 | 456 | 2649589 | 118 | |
More Information
Set Membership: |
|

Cog6
Genomic Map