Assay Details
| Gene Symbol: Cog6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026225.3 | NP_080501.2 | 9 - 10 | 943 | 63 | |
| GenBank mRNA | AB093286.1 | - | 9 - 10 | 915 | 63 | |
| AK016663.1 | - | 9 - 10 | 955 | 63 | ||
| AK031149.1 | - | 9 - 10 | 937 | 63 | ||
| AK045223.1 | - | 9 - 10 | 937 | 63 | ||
| AK048986.1 | - | 9 - 10 | 930 | 63 | ||
| AK153320.1 | - | 9 - 10 | 932 | 63 | ||
| BC025427.1 | - | 9 - 10 | 949 | 2649589 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cog6
Genomic Map