Assay Details
Gene Symbol: Fnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173753.4 | NP_776114.2 | 11 - 12 | 1321 | 103 | |
GenBank mRNA | AK087849.1 | - | 11 - 12 | 1975 | 103 | |
AK122574.1 | - | 11 - 12 | 1269 | 103 | ||
BC079892.1 | - | 11 - 12 | 1311 | 30543296 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
