Assay Details
| Gene Symbol: Kif3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290805.1 | NP_001277734.1 | 13 - 14 | 1953 | 82 | |
| NM_001290806.1 | NP_001277735.1 | 14 - 15 | 1941 | 82 | ||
| NM_008443.4 | NP_032469.2 | 14 - 15 | 1962 | 82 | ||
| GenBank mRNA | AK147078.1 | - | 13 - 14 | 1903 | 82 | |
| AK169425.1 | - | 14 - 15 | 1915 | 82 | ||
| BC052707.1 | - | 14 - 15 | 1928 | 5702415 | 82 | |
| D12645.1 | - | 14 - 15 | 1938 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kif3a
Genomic Map